Learn More
ABclonal Technology PBCV-1 DNA Ligase
SDP
Supplier:  ABclonal Technology RK205136250U
Encompass_Preferred
PBCV-1 DNA Ligase, also known as PBCV DNA Ligase, SplintR(TM) Ligase, or Chorella virus DNA Ligase, is an ATP-dependent DNA ligase that efficiently catalyzes the joining of two adjacent DNA single strands using a complementary RNA strand as a splint. It remains active between 16-37°C. Common applications include the detection of small RNAs such as microRNAs using probe-based methods, utilizing DNA probes to connect and detect specific RNAs, SNP or alternative splicing detection, and RASL-seq analysis.
Catalog No. 50-001-35771