Learn More
ABclonal Technology Mth RNA Ligase
SDP
Catalog No. 5000138405
Encompass_Preferred
Mth RNA Ligase is an ATP-dependent ligase that can efficiently convert 5-phosphorylated single-stranded DNA into 5-adenylated DNA. The kit has been optimized to yield high quantities of adenylylated DNA regardless of whether the 3 end of the single-stranded DNA has been aminated or otherwise modified. Typically, the modification efficiency can reach over 95%.
Catalog No. 50-001-38405 Supplier ABclonal Technology Supplier No. RK2051510RXN
Mth RNA Ligase is an ATP-dependent ligase that can efficiently convert 5-phosphorylated single-stranded DNA into 5-adenylated DNA. The kit has been optimized to yield high quantities of adenylylated DNA regardless of whether the 3 end of the single-stranded DNA has been aminated or otherwise modified. Typically, the modification efficiency can reach over 95%.