Learn More
ABclonal Technology Mth RNA Ligase
SDP
Supplier:  ABclonal Technology RK2051550RXN
Encompass_Preferred
Mth RNA Ligase is an ATP-dependent ligase that can efficiently convert 5-phosphorylated single-stranded DNA into 5-adenylated DNA. The kit has been optimized to yield high quantities of adenylylated DNA regardless of whether the 3 end of the single-stranded DNA has been aminated or otherwise modified. Typically, the modification efficiency can reach over 95%.
Catalog No. 50-001-35180