Learn More
ABclonal Technology DNA Polymerase I, Large (Klenow) Fragment
SDP
Catalog No. 5000137475
Encompass_Preferred
DNA Polymerase I, Large (Klenow) Fragment (about 68 kD) is a proteolytic product of E. coli DNA Polymerase I which retains polymerization and 3 to 5 exonuclease activity, but has lost 5 to 3 exonuclease activity. Klenow retains the polymerization fidelity of the holoenzyme without degrading 5 termini. It is applicable to DNA sequencing by the Sanger dideoxy method, fill-in of 5 overhangs to form blunt ends, removal of 3 overhangs to form blunt ends, second strand cDNA synthesis and second strand synthesis in mutagenesis protocols.
Catalog No. 50-001-37475 Supplier ABclonal Technology Supplier No. RK205251000U
DNA Polymerase I, Large (Klenow) Fragment (about 68 kD) is a proteolytic product of E. coli DNA Polymerase I which retains polymerization and 3 to 5 exonuclease activity, but has lost 5 to 3 exonuclease activity. Klenow retains the polymerization fidelity of the holoenzyme without degrading 5 termini. It is applicable to DNA sequencing by the Sanger dideoxy method, fill-in of 5 overhangs to form blunt ends, removal of 3 overhangs to form blunt ends, second strand cDNA synthesis and second strand synthesis in mutagenesis protocols.