Learn More
ABclonal Technology Klenow Fragment 3'to5' exo-
SDP
Supplier:  ABclonal Technology RK205265000U
Encompass_Preferred
Klenow Fragment (3 to 5 exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5 to 3 exonuclease activity and has mutations (D355A, E357A) which abolish the 3 to 5 exonuclease activity. Klenow Fragment (3 to 5 exo-) is isolated from a recombinant source. It generates probes using random primers and shows moderate strand displacement activity. It can be used in random primer labeling, DNA sequencing by the Sanger dideoxy method, second strand cDNA synthesis and second strand synthesis in mutagenesis protocols.
Catalog No. 50-001-35966