Learn More
ABclonal Technology Uracil-DNA Glycosylase (UDG)
SDP
Catalog No. 5000135802
Encompass_Preferred
E.coli Uracil-DNA Glycosylase (UDG) catalyses the release of free uracil from uracil-containing DNA. UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA, but not from oligomers (6 or fewer bases). It releases uracil from ss- or ds-DNA and is applicable to eliminates PCR carry-over contamination.
Catalog No. 50-001-35802 Supplier ABclonal Technology Supplier No. RK205275000U
E.coli Uracil-DNA Glycosylase (UDG) catalyses the release of free uracil from uracil-containing DNA. UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA, but not from oligomers (6 or fewer bases). It releases uracil from ss- or ds-DNA and is applicable to eliminates PCR carry-over contamination.