Learn More
ABclonal Technology Lambda Exonuclease
SDP
Catalog No. 5000135942
Encompass_Preferred
Lambda Exonuclease digests linear or nicked dsDNA in the 5 to 3 direction. It exhibits the highest cleavage activity towards 5-phosphorylated dsDNA, showing low enzymatic activity toward ssDNA and non-phosphorylated DNA. It remains inactive at DNA nicks but shows limited activity at gaps.
Catalog No. 50-001-35942 Supplier ABclonal Technology Supplier No. RK205291000U
Lambda Exonuclease digests linear or nicked dsDNA in the 5 to 3 direction. It exhibits the highest cleavage activity towards 5-phosphorylated dsDNA, showing low enzymatic activity toward ssDNA and non-phosphorylated DNA. It remains inactive at DNA nicks but shows limited activity at gaps.