Learn More
ABclonal Technology DNA Polymerase I (E. coli)
SDP
Catalog No. 5000135982
Encompass_Preferred
DNA Polymerase I (E coli) is a DNA-dependent DNA polymerase with inherent 3?to 5? and 5?to 3? exonuclease activities. The 5?to 3? exonuclease activity removes nucleotides ahead of the growing DNA chain, allowing nick-translation. It is applicable to nick translation of DNA for obtaining probes with a high specific activity and for second strand synthesis of cDNA.
Catalog No. 50-001-35982 Supplier ABclonal Technology Supplier No. RK205302500U
DNA Polymerase I (E coli) is a DNA-dependent DNA polymerase with inherent 3?to 5? and 5?to 3? exonuclease activities. The 5?to 3? exonuclease activity removes nucleotides ahead of the growing DNA chain, allowing nick-translation. It is applicable to nick translation of DNA for obtaining probes with a high specific activity and for second strand synthesis of cDNA.