Learn More
ABclonal Technology Exonuclease I (E. coli)
SDP
Catalog No. 5000135983
Encompass_Preferred
Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA in the 3 to 5 direction, and does not degrade DNA strands with the 3-OH end blocked by phosphoryl or acetyl groups. Exonuclease I has strict substrate specificity and is incapable of degrading double-stranded DNA and RNA.
Catalog No. 50-001-35983 Supplier ABclonal Technology Supplier No. RK205311500U
Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA in the 3 to 5 direction, and does not degrade DNA strands with the 3-OH end blocked by phosphoryl or acetyl groups. Exonuclease I has strict substrate specificity and is incapable of degrading double-stranded DNA and RNA.