Learn More
ABclonal Technology Exonuclease I (E. coli)
SDP
Supplier:  ABclonal Technology RK2053115000U
Encompass_Preferred
Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA in the 3 to 5 direction, and does not degrade DNA strands with the 3-OH end blocked by phosphoryl or acetyl groups. Exonuclease I has strict substrate specificity and is incapable of degrading double-stranded DNA and RNA.
Catalog No. 50-001-37447