Learn More
ABclonal Technology RNase H
SDP
Supplier:  ABclonal Technology RK205351250U
Encompass_Preferred
RNase H (Ribonuclease H) is an endoribonuclease that specifically hydrolyzes the phosphodiester bonds of RNA which is hybridized to DNA. This enzyme does not digest single or double-stranded DNA.
Catalog No. 50-001-35943