Learn More
ABclonal Technology T4 DNA Polymerase (5,000 U/mL)
SDP
Catalog No. 5000138645
Encompass_Preferred
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5to3 direction and requires the presence of template and primer. This enzyme has a 3to5 exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5to3 exonuclease function. It is applicable to 3 overhang removal to form blunt ends, 5 overhang fill-in to form blunt ends, single strand deletion subcloning, second strand synthesis in site-directed mutagenesis and probe labeling using replacement synthesis.
Catalog No. 50-001-38645 Supplier ABclonal Technology Supplier No. RK20539200U
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5to3 direction and requires the presence of template and primer. This enzyme has a 3to5 exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5to3 exonuclease function. It is applicable to 3 overhang removal to form blunt ends, 5 overhang fill-in to form blunt ends, single strand deletion subcloning, second strand synthesis in site-directed mutagenesis and probe labeling using replacement synthesis.