Learn More
ABclonal Technology Heat-labile UDG (1,000 U/mL)
SDP
Supplier:  ABclonal Technology RK20543500U
Encompass_Preferred
Heat-labile UDG was derived from psychrophilic marine bacteria. It can catalyze the release of free uracil from uracil (U)-containing DNA and is inactive against RNA. This product is sensitive to high temperature, and it can cause irreversible inactivation of the enzyme above 50℃, so it is suitable for PCR qPCR RT-PCR RT-qPCR system.
Catalog No. 50-001-35807