Learn More
ABclonal Technology DNase I , RNase-free (5,000 U/mL)
SDP
Catalog No. 5000136010
Encompass_Preferred
DNase I (deoxyribonuclease, RNase-free ) is an endonuclease that nonspecifically cleaves DNA to release di-, tri-, and oligonucleotide products with 5phosphorylated and 3?-hydroxylated ends. The activity of DNase I depends on Ca2+ and also be activated bydivalent metal ions Mg2+, Mn2+, etc.DNase I act on various DNAs such as single and double-stranded DNA, RNA:DNA hybrids.
Catalog No. 50-001-36010 Supplier ABclonal Technology Supplier No. RK205495000U
DNase I (deoxyribonuclease, RNase-free ) is an endonuclease that nonspecifically cleaves DNA to release di-, tri-, and oligonucleotide products with 5phosphorylated and 3?-hydroxylated ends. The activity of DNase I depends on Ca2+ and also be activated bydivalent metal ions Mg2+, Mn2+, etc.DNase I act on various DNAs such as single and double-stranded DNA, RNA:DNA hybrids.