Learn More
ABclonal Technology T7 Endonuclease I
SDP
Catalog No. 5000135808
Encompass_Preferred
T7 Endonuclease I (T7 Endo I, T7EI),can recognize and cleave non-perfectly mismatched DNA, cruciform DNA structures, holliday structures or junctions and Heteroduplex DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5? end to the mismatch.
Catalog No. 50-001-35808 Supplier ABclonal Technology Supplier No. RK205541250U
T7 Endonuclease I (T7 Endo I, T7EI),can recognize and cleave non-perfectly mismatched DNA, cruciform DNA structures, holliday structures or junctions and Heteroduplex DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5? end to the mismatch.