Learn More
ABclonal Technology T7 Endonuclease I
SDP
Supplier:  ABclonal Technology RK20554250U
Encompass_Preferred
T7 Endonuclease I (T7 Endo I, T7EI),can recognize and cleave non-perfectly mismatched DNA, cruciform DNA structures, holliday structures or junctions and Heteroduplex DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5? end to the mismatch.
Catalog No. 50-001-36011