Learn More
ABclonal Technology T7 DNA Ligase
SDP
Supplier:  ABclonal Technology RK20555100000U
Encompass_Preferred
T7 DNA Ligase is an ATP-dependent ds DNA ligase from bacteriophage T7. It can catalyze the formation of a phosphodiester bond between adjacent 5 phosphate and 3 hydroxyl termini in double-stranded DNA. T7 DNA ligase can catalyze ligation of cohesive ends and repair nicks, but not blunt end ligation. This makes a good choice for applications in which blunt and cohesive ends of DNA are present but only the cohesive ends are to be joined .
Catalog No. 50-001-35809