Learn More
ABclonal Technology Exonuclease VII
SDP
Supplier:  ABclonal Technology RK205631000U
Encompass_Preferred
Exonuclease VII (Exo VII) can cleave single-stranded DNA in both 5-3 and 3-5 directions and is inactive against linear or circular double-stranded DNA. When PCR requires stepwise completion to obtain the final target product, this enzyme can be used to remove residual oligonucleotide primers left during intermediate stages of PCR, eliminating interference with subsequent use of different primers. The enzymes digestion of single-stranded DNA is independent of metal ions. This product is a highly purified protein obtained through the recombinant expression of the Exonuclease VII gene (XseA and XseB).
Catalog No. 50-001-38418