Learn More
ABclonal Technology PNGase F, Recombinant (Expressed in E.coli)
SDP
Catalog No. 5000135215
Encompass_Preferred
Peptide N-glycosidase F (PNGase F) is a glycosylpeptidase with a theoretical molecular weight of 35.9 kD that can cleave asparagine-linked high mannose, hybrid, and complex oligosaccharides from glycoproteins. PNGase F cleaves the amide bond between innermost N-acetylglucosamine (GlcNAc) and asparagine residues of the glycoprotein, converting asparagine to aspartic acid in the process. This product is commonly used for the deglycosylation of antibodies and glycoproteins.
Catalog No. 50-001-35215 Supplier ABclonal Technology Supplier No. RK2056520000U
Peptide N-glycosidase F (PNGase F) is a glycosylpeptidase with a theoretical molecular weight of 35.9 kD that can cleave asparagine-linked high mannose, hybrid, and complex oligosaccharides from glycoproteins. PNGase F cleaves the amide bond between innermost N-acetylglucosamine (GlcNAc) and asparagine residues of the glycoprotein, converting asparagine to aspartic acid in the process. This product is commonly used for the deglycosylation of antibodies and glycoproteins.