Learn More
ABclonal Technology Bsu DNA Polymerase
SDP
Catalog No. 5000138419
Encompass_Preferred
Bsu DNA Polymerase, Large Fragment retains the 5?to 3? polymerase activity of DNA polymerase I but lacks the 5?to 3? exonuclease activity. This large fragment also lacks 3?to 5? exonuclease activity. Bsu DNA polymerase has strand displacement activity and is commonly used in recombinase polymerase amplification (RPA).
Catalog No. 50-001-38419 Supplier ABclonal Technology Supplier No. RK205661000U
Bsu DNA Polymerase, Large Fragment retains the 5?to 3? polymerase activity of DNA polymerase I but lacks the 5?to 3? exonuclease activity. This large fragment also lacks 3?to 5? exonuclease activity. Bsu DNA polymerase has strand displacement activity and is commonly used in recombinase polymerase amplification (RPA).