Learn More
ABclonal Technology RK20567-150-ug T4 UvsX Recombi
SDP
Catalog No. 5000142629
Encompass_Preferred
T4 UvsX Recombinase is homologous to the RecA/Rad51 family The RecA/Rad51 family of recombinases plays an important role in gene recombination DNA repair and replication with pairing and strand replacement activities T4 UvsX Recombinase can bind with primers to form a complex to search for homologous sequences in double stranded DNA and perform strand displacement reactions It can work together with T4 UvsY protein single stranded DNA binding protein Bsu polymerase etc to complete isothermal amplification (RPA) of recombinase polymerase
Catalog No. 50-001-42629 Supplier ABclonal Technology Supplier No. RM20548150MG
T4 UvsX Recombinase is homologous to the RecA/Rad51 family The RecA/Rad51 family of recombinases plays an important role in gene recombination DNA repair and replication with pairing and strand replacement activities T4 UvsX Recombinase can bind with primers to form a complex to search for homologous sequences in double stranded DNA and perform strand displacement reactions It can work together with T4 UvsY protein single stranded DNA binding protein Bsu polymerase etc to complete isothermal amplification (RPA) of recombinase polymerase