Learn More
ABclonal Technology Endonuclease III (Nth)
SDP
Catalog No. 5000137450
Encompass_Preferred
Endonuclease III (Nth) protein acts as both N-glycosylase and a AP-lyase.The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating a basic (AP site). The AP-lyase activity of the enzyme cleaves 3? to the AP site leaving a 5? phosphate and a 3? ring opened sugar.
Catalog No. 50-001-37450 Supplier ABclonal Technology Supplier No. RK205691000U
Endonuclease III (Nth) protein acts as both N-glycosylase and a AP-lyase.The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating a basic (AP site). The AP-lyase activity of the enzyme cleaves 3? to the AP site leaving a 5? phosphate and a 3? ring opened sugar.