Learn More
ABclonal Technology T7 Exonuclease
SDP
Catalog No. 5000137451
Encompass_Preferred
T7 Exonuclease is a double-stranded DNA-specific exonuclease that degrades linear or nicked double-stranded DNA in the 5′to 3′direction. The enzyme does not digest supercoiled dsDNA.
Catalog No. 50-001-37451 Supplier ABclonal Technology Supplier No. RK205741000U
T7 Exonuclease is a double-stranded DNA-specific exonuclease that degrades linear or nicked double-stranded DNA in the 5′to 3′direction. The enzyme does not digest supercoiled dsDNA.