Learn More
ABclonal Technology T7 Exonuclease
SDP
Supplier:  ABclonal Technology RK205745000U
Encompass_Preferred
T7 Exonuclease is a double-stranded DNA-specific exonuclease that degrades linear or nicked double-stranded DNA in the 5′to 3′direction. The enzyme does not digest supercoiled dsDNA.
Catalog No. 50-001-36004