Learn More
ABclonal Technology 5' Deadenylase
SDP
Catalog No. 5000136003
Encompass_Preferred
The 5 Deadenylase derived from yeast can remove adenosine residues from the 5-end of DNA and RNA, retaining the 5-end phosphate group. It can cleave AppppA to produce ATP and AMP.
Catalog No. 50-001-36003 Supplier ABclonal Technology Supplier No. RK205802500U
The 5 Deadenylase derived from yeast can remove adenosine residues from the 5-end of DNA and RNA, retaining the 5-end phosphate group. It can cleave AppppA to produce ATP and AMP.