Learn More
ABclonal Technology RK20589-500-ug Thermostable SS
SDP
Catalog No. 5000135183
Encompass_Preferred
Thermostable SSB (Thermostable Single Stranded Binding Protein) is a single-stranded DNA-binding protein isolated from extremely heat-tolerant microorganisms It remains fully activity after 60 min of incubation at 95 C Due to its high thermal stability Thermostable SSB can be used in experiments that require high-temperature reaction conditions such as to increase PCR yield and specificity as well as to increase isothermal amplification yields and enhance the efficiency of reverse transcription in RT-PCR
Catalog No. 50-001-35183 Supplier ABclonal Technology Supplier No. RK20589500MG
Thermostable SSB (Thermostable Single Stranded Binding Protein) is a single-stranded DNA-binding protein isolated from extremely heat-tolerant microorganisms It remains fully activity after 60 min of incubation at 95 C Due to its high thermal stability Thermostable SSB can be used in experiments that require high-temperature reaction conditions such as to increase PCR yield and specificity as well as to increase isothermal amplification yields and enhance the efficiency of reverse transcription in RT-PCR