Learn More
ABclonal Technology RK20590-100-ug E.coli SSB
SDP
Catalog No. 5000135184
Encompass_Preferred
E coli SSB (single-stranded binding) protein is a single-stranded DNA (ssDNA) binding protein essential for E coli DNA replication and repair The tetramer formed by the SSB protein specifically binds 8-16 nucleotides thereby protecting ssDNA from nuclease degradation In vitro experiments have shown that this protein can stabilize single-stranded regions of DNA thereby relaxing double-stranded regions which enhances the activity of DNA polymerase
Catalog No. 50-001-35184 Supplier ABclonal Technology Supplier No. RK20590100MG
E coli SSB (single-stranded binding) protein is a single-stranded DNA (ssDNA) binding protein essential for E coli DNA replication and repair The tetramer formed by the SSB protein specifically binds 8-16 nucleotides thereby protecting ssDNA from nuclease degradation In vitro experiments have shown that this protein can stabilize single-stranded regions of DNA thereby relaxing double-stranded regions which enhances the activity of DNA polymerase