Learn More
ABclonal Technology E. coli Poly(A) Polymerase
SDP
Supplier:  ABclonal Technology RK20591100U
Encompass_Preferred
Poly(A) polymerase catalyzes the addition of AMP, converted from ATP, to the 3 end of RNA in a template-independent manner.
Catalog No. 50-001-36492