Learn More
ABclonal Technology E. coli Poly(A) Polymerase
SDP
Catalog No. 5000135944
Encompass_Preferred
Poly(A) polymerase catalyzes the addition of AMP, converted from ATP, to the 3 end of RNA in a template-independent manner.
Catalog No. 50-001-35944 Supplier ABclonal Technology Supplier No. RK20591500U
Poly(A) polymerase catalyzes the addition of AMP, converted from ATP, to the 3 end of RNA in a template-independent manner.