Learn More
ABclonal Technology Taq DNA Polymerase (Mg2+ Plus Buffer)
SDP
Supplier:  ABclonal Technology RK206005000U
Encompass_Preferred
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5?to 3? polymerase activity and a 5? flap endonuclease activity. Taq DNA Polymerase was isolated from a recombinant source and offers robust and reliable reactions. It tolerates a wide range of templates and incorporates dUTP, dITP, and fluorescently-labeled nucleotides.
Catalog No. 50-001-37452