Learn More
ABclonal Technology Taq 2X PCR Mix V2
SDP
Catalog No. 5000135899
Encompass_Preferred
Taq 2X PCR Mix V2 is an optimized PCR premix solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCI and other stabilizers. User only need to add template and primer to complete experiment. It is ideally suited to routine PCR applications from various templates including pure DNA solutions, bacterial colony/culture, and cDNA products. It can amplify up to 5 kb DNA from different sources genomic DNA. It is applicable to PCR reaction, colony PCR identification, rough sample amplification.
Catalog No. 50-001-35899 Supplier ABclonal Technology Supplier No. RK206071ML
Taq 2X PCR Mix V2 is an optimized PCR premix solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCI and other stabilizers. User only need to add template and primer to complete experiment. It is ideally suited to routine PCR applications from various templates including pure DNA solutions, bacterial colony/culture, and cDNA products. It can amplify up to 5 kb DNA from different sources genomic DNA. It is applicable to PCR reaction, colony PCR identification, rough sample amplification.