Learn More
ABclonal Technology Taq 2X PCR Mix with Dye V2
SDP
Catalog No. 5000136628
Encompass_Preferred
Taq 2X PCR Mix with Dye V2 is an optimized PCR premixed solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCI, Bromophenol blue tracking dye, and other stabilizers. User only need to add template and primer to complete experiment. The amplified products were directly used for agarose gel electrophoresis. It is ideally suited to routine PCR applications on various templates including pure DNA solutions, bacterial colony/culture, and cDNA products. It can amplify 5 kb DNA from different sources genomic DNA. It is applicable to PCR reaction, colony PCR identification, rough sample amplification.
Catalog No. 50-001-36628 Supplier ABclonal Technology Supplier No. RK206081ML
Taq 2X PCR Mix with Dye V2 is an optimized PCR premixed solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCI, Bromophenol blue tracking dye, and other stabilizers. User only need to add template and primer to complete experiment. The amplified products were directly used for agarose gel electrophoresis. It is ideally suited to routine PCR applications on various templates including pure DNA solutions, bacterial colony/culture, and cDNA products. It can amplify 5 kb DNA from different sources genomic DNA. It is applicable to PCR reaction, colony PCR identification, rough sample amplification.