Learn More
ABclonal Technology Gloria Nova HS PCR Kit
SDP
Supplier:  ABclonal Technology RK2071450RXN
Encompass_Preferred
Gloria Nova HS DNA Polymerase is a high-fidelity engineered enzyme rivaling Pyrococcus furiosus DNA polymerase. Featuring a synthesis-enhancing domain, it delivers fast extension speeds and ultra-high fidelity (10–15× Taq, 6× P. furiosus), aided by strong 3’to5’ proofreading. Its antibody-mediated hot-start prevents nonspecific amplification. The kit includes 5X HF Buffer (general high-fidelity PCR) and 2.5X GC Buffer (GC-rich/problematic templates), ensuring robust performance across diverse applications.
Catalog No. 50-001-36008