Learn More
ABclonal Technology PowerPol 2X PCR Mix
SDP
Catalog No. 5000138644
Encompass_Preferred
PowerPol 2X PCR Mix is an optimized premix containing DNA polymerase, dNTPs, MgCl2, KCI and other stabilizers. This product is suitable for conventional PCR amplification. The template can be purified DNA, bacterial colonies/bacteria liquid, crude extract or cDNA, etc. This product can use complex genomic DNA as a template to amplify a target fragment of 5 kb in length or a simple template such as lambda DNA to amplify a target fragment of 10 kb in length. It is suitable for applications such as PCR reaction, colony PCR identification, vector construction and so on. etc.
Catalog No. 50-001-38644 Supplier ABclonal Technology Supplier No. RK20718100ML
PowerPol 2X PCR Mix is an optimized premix containing DNA polymerase, dNTPs, MgCl2, KCI and other stabilizers. This product is suitable for conventional PCR amplification. The template can be purified DNA, bacterial colonies/bacteria liquid, crude extract or cDNA, etc. This product can use complex genomic DNA as a template to amplify a target fragment of 5 kb in length or a simple template such as lambda DNA to amplify a target fragment of 10 kb in length. It is suitable for applications such as PCR reaction, colony PCR identification, vector construction and so on. etc.