Learn More
ABclonal Technology TruePol 2X PCR Mix for Microbiome
SDP
Catalog No. 5000137453
Encompass_Preferred
Truepol 2X PCR Mix for Microbiome is developed for microbial diversity detection, having high amplification efficiency for various types of samples. Certain specific components mixted in this reagent enhance inhibitor tolerance and hence to improve the sensitivity. Even there is large amount of host DNA as interference in samples, Truepol 2X PCR Mix for Microbiome ensures good amplification efficiency, significantly improving the success of microbial detection rate. Truepol 2X PCR Mix for Microbiome can be used for dealing with general and complex samples, such as human tissue, soil, feces, urine, air filter membrane, sand, etc.
Catalog No. 50-001-37453 Supplier ABclonal Technology Supplier No. RK207201ML
Truepol 2X PCR Mix for Microbiome is developed for microbial diversity detection, having high amplification efficiency for various types of samples. Certain specific components mixted in this reagent enhance inhibitor tolerance and hence to improve the sensitivity. Even there is large amount of host DNA as interference in samples, Truepol 2X PCR Mix for Microbiome ensures good amplification efficiency, significantly improving the success of microbial detection rate. Truepol 2X PCR Mix for Microbiome can be used for dealing with general and complex samples, such as human tissue, soil, feces, urine, air filter membrane, sand, etc.