Learn More
ABclonal Technology PCR Enhancer for Microbiome
Supplier: ABclonal Technology RK207251ML
For certain difficult samples that cannot be effectively amplified or generate non-specific amplification products, using RK20725 PCR Enhancer for Microbiome with RK20720 TruePol 2X PCR Mix for Microbiome improve the specificity and success rate of amplification. RK20720 Truepol 2X PCR Mix for Microbiome is suitable for amplification of general and complex sample source templates, such as human tissue samples, soil samples, fecal samples, air filter membrane samples, sand samples, etc.