Learn More
ABclonal Technology PowerPol 2X PCR Mix with Dye V2
SDP
Supplier:  ABclonal Technology RK207295ML
Encompass_Preferred
PowerPol 2X PCR Mix with Dye V2 is an optimized premix with amplification rates up to 10 s/kb to save PCR reaction time. Reagents include DNA polymerases, dNTPs, MgCl2, KCI, and other stabilizers. It only needs to add primers and templates to perform amplification. This product contains loading buffer, PCR products can be directly loaded for electrophoresis. PowerPol 2X PCR Mix with Dye V2 can use complex genomic DNA as a template to amplify a 5 kb target fragment or a simple template such as lambda DNA to amplify a 10 kb target fragment. It is suitable for colony PCR, genotype identification, vector construction, and other applications.
Catalog No. 50-001-35805