Learn More
ABclonal Technology Gloria Nova HS 2X Master Mix with Dye V2
SDP
Supplier:  ABclonal Technology RK207305ML
Encompass_Preferred
This product utilizes antibody-blocked high-fidelity DNA polymerase, enabling high-specificity hot-start PCR. It has approximately 120× the fidelity of Taq polymerase, making it ideal for molecular cloning and effective for various templates such as animal, plant, and cDNA samples.
Catalog No. 50-001-35904