Learn More
ABclonal Technology Gloria Nova HS 2X Master Mix V2
SDP
Catalog No. 5000137456
Encompass_Preferred
This product utilizes antibody-blocked high-fidelity DNA polymerase, enabling high-specificity hot-start PCR. It has approximately 120× the fidelity of Taq polymerase, making it ideal for molecular cloning and effective for various templates such as animal, plant, and cDNA samples.
Catalog No. 50-001-37456 Supplier ABclonal Technology Supplier No. RK207315ML
This product utilizes antibody-blocked high-fidelity DNA polymerase, enabling high-specificity hot-start PCR. It has approximately 120× the fidelity of Taq polymerase, making it ideal for molecular cloning and effective for various templates such as animal, plant, and cDNA samples.