Learn More
ABclonal Technology Mouse Tissue Direct PCR Kit
Supplier: ABclonal Technology RK20736500RXN
Mouse Tissue Direct PCR Kit is specially designed for the rapid genotyping of mouse, which contains Tissue Lysis Buffer II and Powerpol HS 2X PCR Mix for Mouse Genotyping for DNA release and PCR amplification. This kit can be used for the rapid extraction of genomic DNA from mouse tails, ears, toes, and other tissues. The extracted genomic DNA can be used directly as template for PCR amplification, which greatly shortens the experimental time. Tissue Lysis Buffer II in this kit already contains proteinase K, which does not require additional preparation. Powerpol HS 2X PCR Mix for Mouse Genotyping in this kit only needs to add primers and templates to perform amplification, and the reagent contains loading buffer, PCR products can be directly loaded for electrophoresis.