Learn More
ABclonal Technology Powerpol HS 2X PCR Mix for Mouse Genotyping
SDP
Supplier:  ABclonal Technology RK207371.250ML
Encompass_Preferred
Powerpol HS 2X PCR Mix for Mouse Genotyping is one of kit components of Mouse Tissue Direct PCR Kit (ABclonal RK20736), which was used for extra needs when user are dealing with multi-primer amplification and consuming such mix in a volume larger than a regular kit size form. Powerpol HS 2X PCR Mix for Mouse Genotyping only needs to add primers and templates to perform amplification, and the reagent contains loading buffer, PCR products can be directly loaded for electrophoresis.
Catalog No. 50-001-35917