Learn More
ABclonal Technology Ruby Bst DNA Polymerase
SDP
Supplier:  ABclonal Technology RK210051600U
Encompass_Preferred
Ruby Bst DNA Polymerase is an in silico designed homologue of Geobacillus stearothermophilus DNA Polymerase I(large fragment). Ruby Bst DNA Polymerase has 5?to3? DNA polymerase activity and strong strand displacement activity but lacks 5?to3? exonuclease activity. Ruby Bst DNA Polymerase displays improved amplification speed, yield, salt tolerance and thermo stability compared with wild-type Bst DNA Polymerase, Large Fragment.
Catalog No. 50-001-35812