Learn More
ABclonal Technology Pilot phi29 DNA Polymerase
SDP
Catalog No. 5000135837
Encompass_Preferred
Pilot phi29 DNA Polymerase differs from Phi29 DNA Polymerase by significantly enhancing protein thermal stability, reaction speed, and salt tolerance while retaining all the advantages of the latter. Pilot phi29 DNA Polymerase exhibits strong multiple strand displacement, continuous synthesis characteristics, and 3 -5 exonuclease activity.
Catalog No. 50-001-35837 Supplier ABclonal Technology Supplier No. RK210061250U
Pilot phi29 DNA Polymerase differs from Phi29 DNA Polymerase by significantly enhancing protein thermal stability, reaction speed, and salt tolerance while retaining all the advantages of the latter. Pilot phi29 DNA Polymerase exhibits strong multiple strand displacement, continuous synthesis characteristics, and 3 -5 exonuclease activity.