Learn More
ABclonal Technology Pilot phi29 DNA Polymerase
SDP
Catalog No. 5000135838
Encompass_Preferred
Pilot phi29 DNA Polymerase differs from Phi29 DNA Polymerase by significantly enhancing protein thermal stability, reaction speed, and salt tolerance while retaining all the advantages of the latter. Pilot phi29 DNA Polymerase exhibits strong multiple strand displacement, continuous synthesis characteristics, and 3 -5 exonuclease activity.
Catalog No. 50-001-35838 Supplier ABclonal Technology Supplier No. RK21006250U
Pilot phi29 DNA Polymerase differs from Phi29 DNA Polymerase by significantly enhancing protein thermal stability, reaction speed, and salt tolerance while retaining all the advantages of the latter. Pilot phi29 DNA Polymerase exhibits strong multiple strand displacement, continuous synthesis characteristics, and 3 -5 exonuclease activity.