Learn More
ABclonal Technology XhoI
SDP
Catalog No. 5000136031
Encompass_Preferred
5...C/TCGAG...3 3...GAGCT/C...5 Isoschizomers: PaeR7I Sfr274I SlaI
Catalog No. 50-001-36031 Supplier ABclonal Technology Supplier No. RK211005000U
5...C/TCGAG...3 3...GAGCT/C...5 Isoschizomers: PaeR7I Sfr274I SlaI