Learn More
ABclonal Technology BamHI
SDP
Catalog No. 5000136012
Encompass_Preferred
5...G/GATCC...3 3...CCTAG/G...5
Catalog No. 50-001-36012 Supplier ABclonal Technology Supplier No. RK2110150000U
5...G/GATCC...3 3...CCTAG/G...5