Learn More
ABclonal Technology EcoRI
SDP
Catalog No. 5000138421
Encompass_Preferred
5...G/ A A T T C...3 3...C T T A A/ G...5
Catalog No. 50-001-38421 Supplier ABclonal Technology Supplier No. RK2110210000U
5...G/ A A T T C...3 3...C T T A A/ G...5