Learn More
ABclonal Technology NotI
SDP
Catalog No. 5000136498
Encompass_Preferred
5...GC/GGCCGC...3 3...CGCCGG/CG...5 Isoschizomers: CciNI
Catalog No. 50-001-36498 Supplier ABclonal Technology Supplier No. RK21103500U
5...GC/GGCCGC...3 3...CGCCGG/CG...5 Isoschizomers: CciNI