Learn More
ABclonal Technology KpnI
SDP
Catalog No. 5000135959
Encompass_Preferred
5...GGTAC/C...3 3...C/CATGG...5
Catalog No. 50-001-35959 Supplier ABclonal Technology Supplier No. RK2110420000U
5...GGTAC/C...3 3...C/CATGG...5