Learn More
ABclonal Technology SfiI
SDP
Catalog No. 5000136032
Encompass_Preferred
5…GGCCNNNN/NGGCC...3 3…CCGGN/NNNNCCGG...5
Catalog No. 50-001-36032 Supplier ABclonal Technology Supplier No. RK2110515000U
5…GGCCNNNN/NGGCC...3 3…CCGGN/NNNNCCGG...5